Isoamyl alcohol 99,0, AnalaR NORMAPUR® zA

Supplier: VWR Chemicals
Warning

Synonyms: Isopentyl alcohol, iso-Pentanol, 3-Methyl-1-butanol, 3-Methylbutanol

VWRC20798.295EA 103 SGD
VWRC20798.295
Isoamyl alcohol 99,0, AnalaR NORMAPUR® zA
Isoamyl alcohol
Formula: (CH₃)₂CHCH₂CH₂OH
MW: 88.15 g/mol
Boiling Pt: 130
Melting Pt: –117 °C
Density: 0.812 g/cm³ (20 °C)
Flash Pt: 42
Storage Temperature: Ambient
MDL Number: MFCD00002934
CAS Number: 123-51-3
EINECS: 204-633-5
UN: 1105
ADR: 3 III
Merck Index: 13,05214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd7b89cc1890b5d4b2083195c30c0d1047c32e9a&vid=170c60e26c7b4edab6e0686f91d04e676da16e6d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (calculated as mixture of isomers) Min. 99.0 %
Acidity Max. 0.008 meq/g
Alkalinity Max. 0.0008 meq/g
Boiling point 131 - 132.5 °C
Colouration Max. 10 APHA
Density (20/4) 0.808 - 0.812
Acidity and esters (as C7H14O2) Max. 0.17 %
Evaporation residue Max. 10 ppm
Furfuraldehyde Max. 1 ppm
Pentan-1-ol Max. 0.5 %
Peroxides (as H2O2) Max. 10 ppm
Water Max. 0.2 %
Al (Aluminium) Max. 0.5 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.1 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.02 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.1 ppm
Sn (Tin) Max. 0.1 ppm
Zn (Zinc) Max. 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR