Benzyl alcohol ≥99.0% analytical reagent

Supplier: VWR Chemicals
Warning

Synonyms: Benzenemethanol, Phenylmethanol, Phenylcarbinol

VWRC20807.297EA 95 SGD
VWRC20807.297 VWRC20807.322
Benzyl alcohol ≥99.0% analytical reagent
Benzyl alcohol
Formula: C₆H₅CH₂OH
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: –15,3 °C
Density: 1,05 g/cm³ (20 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS Number: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.0 %
Acidity Max. 0.0008 meq/g
Boiling point 204 - 208 °C
Density (20/4) 1.043 - 1.046
Peroxide value Max. 10
Benzaldehyde Max. 0.1 %
Water Max. 0.1 %
Cl (Chloride) Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR