Aluminium potassium sulphate dodecahydrate 97, TECHNICAL

Supplier: VWR Chemicals

Synonyms: Aluminium Potassium sulphate, 12-Hydrate, Aluminum potassium sulphate dodecahydrate, Aluminium potassium bis(sulphate) dodecahydrate, Potassium alum, Alum potassium

VWRC21098.293EA 41.6 SGD
VWRC21098.293 VWRC21098.362
Aluminium potassium sulphate dodecahydrate 97, TECHNICAL
Aluminium potassium sulphate dodecahydrate
Formula: AlK(SO₄)₂·12H₂O
MW: 474.39 g/mol
Melting Pt: 92.5 °C
Density: 1.72 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149143
CAS Number: 7784-24-9
EINECS: 233-141-3
REACH: 01-2119960162-44
Merck Index: 12,00373

Order Now

Specification Test Results

Assay Min. 97 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR