Ammonium hydrogen carbonate ≥97%, GPR RECTAPUR®

Supplier: VWR Chemicals
Warning

VWRC21218.298EA 72.5 SGD
VWRC21218.298
Ammonium hydrogen carbonate ≥97%, GPR RECTAPUR®
Ammonium hydrogen carbonate
Formula: NH₄HCO₃
Melting Pt: 35 °C
Density: 1,57 g/cm³ (20 °C)
MDL Number: MFCD00012138
CAS Number: 1066-33-7
EINECS: 213-911-5
Merck Index: 13,00497

Order Now

Specification Test Results

Assay Min. 97 %
Heavy metals (as Pb) Max. 20 ppm
Ignition residue (SO4) Max. 0.05 %
Fe (Iron) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR