Ammonium heptamolybdate tetrahydrate, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent

Supplier: VWR Chemicals
Warning

Synonyms: Hexaammonium heptamolybdate tetrahydrate, Ammonium molybdate(VI) tetrahydrate, Ammonium molybdate tetrahydrate, Hexammonium heptamolybdate 4-hydrate, Molybdic acid ammonium salt tetrahydrate

VWRC21276.298EA 246 SGD
VWRC21276.298 VWRC21276.185 VWRC21276.260 VWRC21276.460
Ammonium heptamolybdate tetrahydrate, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Ammonium heptamolybdate tetrahydrate
Formula: (NH₄)₆Mo₇O₂₄·4H₂O
Density: 2,498 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167059
CAS Number: 12054-85-2
EINECS: 234-722-4
Merck Index: 13,00536

Order Now

Specification Test Results

Assay (calculated as MoO3) 81.0 - 83.0 %
Heavy metals (as Pb) Max. 10 ppm
Insolubility in water Max. 50 ppm
Mg and alkaline earth metals Max. 0.02 %
Cl (Chloride) Max. 5 ppm
NO3 (Nitrate) Max. 0.05 %
PO4 + AsO4 + SiO4 (as PO4) Max. 5 ppm
PO4 + AsO4 + SiO4 (as SiO2) Max. 10 ppm
SO4 (Sulphate) Max. 50 ppm
Cu (Copper) Max. 10 ppm
Fe (Iron) Max. 5 ppm
K (Potassium) Max. 100 ppm
Mg (Magnesium) Max. 50 ppm
Na (Sodium) Max. 100 ppm
Pb (Lead) Max. 10 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR