Ammonium peroxodisulphate (APS) ≥97.5%, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: Peroxidisulphuric acid diammonium salt, Ammonium persulphate

VWRC21299.366EA 166 SGD
VWRC21299.366
Ammonium peroxodisulphate (APS) ≥97.5%, GPR RECTAPUR®
Ammonium peroxodisulphate (APS)
Formula: (NH₄)₂S₂O₈
MW: 228,2 g/mol
Melting Pt: 120 °C
Density: 1,98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003390
CAS Number: 7727-54-0
EINECS: 231-786-5
UN: 1444
ADR: 5.1,III
Merck Index: 13,00545
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82a373f21d34e63dc94c0598f3f33ba6bbfdb2b&vid=2b6d196dbf07ce4dd528ac7b4f8456678accd3ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 97.5 %
Heavy metals (as Pb) Max. 50 ppm
Cl (Chloride) Max. 20 ppm
Fe (Iron) Max. 50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR