Ammonium sulphate ≥99%, GPR RECTAPUR®

Supplier: VWR Chemicals

VWRC21332.293EA 39.9 SGD
VWRC21332.293 VWRC21332.362 VWRC21332.460
Ammonium sulphate ≥99%, GPR RECTAPUR®
Ammonium sulphate
Formula: (NH₄)₂SO₄
Melting Pt: 336…339 °C
Density: 1,78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Order Now

Specification Test Results

Assay Min. 99 %
Heavy metals (as Pb) Max. 10 ppm
Ignition residue (SO4) Max. 0.02 %
Cl (Chloride) Max. 20 ppm
NO3 (Nitrate) Max. 50 ppm
Fe (Iron) Max. 10 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR