Antimony(III) chloride ≥99.0%, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals
Danger

Synonyms: Antimony trichloride

VWRC21470.186EA 109 SGD
VWRC21470.186
Antimony(III) chloride ≥99.0%, AnalaR NORMAPUR® analytical reagent
Antimony (III) chloride
Formula: SbCl₃
MW: 228.12 g/mol
Boiling Pt: 223.5 °C (1013 hPa)
Melting Pt: 73
Density: 3.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011212
CAS Number: 10025-91-9
EINECS: 233-047-2
UN: 1733
ADR: 8 II
Merck Index: 13,00713

Order Now

Specification Test Results

Assay Min. 99.0 %
Insolubility in chloroform Max. 0.01 %
Insolubility in HCl Max. 50 ppm
SO4 (Sulphate) Max. 50 ppm
Ca (Calcium) Max. 50 ppm
Cu (Copper) Max. 5 ppm
Fe (Iron) Max. 20 ppm
K (Potassium) Max. 0.01 %
Na (Sodium) Max. 50 ppm
Pb (Lead) Max. 30 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR