Silver sulphate ≥98%, TECHNICAL

Supplier: VWR Chemicals
Danger

VWRC21589.153EA 357 SGD
VWRC21589.153 VWRC21589.233
Silver sulphate ≥98%, TECHNICAL
Silver sulphate
Formula: Ag₂SO₄
Boiling Pt: 1085 °C (1013 hPa)
Melting Pt: 655 °C
Density: 5,45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003407
CAS Number: 10294-26-5
EINECS: 233-653-7
UN: 3077
ADR: 9,III
Merck Index: 13,08603
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd7afaa8e9ae24bf040d40aed0c93763ce897080&vid=ae7b50634b53507047974c2157e5e38c2eb90a20&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR