Silver sulphate 98,5, AnalaR NORMAPUR® zA

Supplier: VWR Chemicals
Danger

Synonyms: Disilver sulphate

VWRC21592.154EA 234 SGD
VWRC21592.154 VWRC21592.234
Silver sulphate 98,5, AnalaR NORMAPUR® zA
Silver sulphate
Formula: Ag₂SO₄
MW: 311.8 g/mol
Boiling Pt: 1085
Melting Pt: 655
Density: 5.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003407
CAS Number: 10294-26-5
EINECS: 233-653-7
UN: 3077
ADR: 9 III
Merck Index: 13,08603
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd7afaa8e9ae24bf040d40aed0c93763ce897080&vid=ae7b50634b53507047974c2157e5e38c2eb90a20&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 98.5 %
Solution in water Passes test
Foreign heavy metals (as Pb) Max. 20 ppm
Insolubility in water Max. 0.02 %
Substances not precipitated by HCl Max. 0.03 %
Cl (Chloride) Max. 20 ppm
Cu (Copper) Max. 5 ppm
Fe (Iron) Max. 10 ppm
Pb (Lead) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR