Barium chloride dihydrate ≥98%, TECHNICAL

Supplier: VWR Chemicals
Danger

Synonyms: Barium dichloride

VWRC21709.364EA 317 SGD
VWRC21709.364
Barium chloride dihydrate ≥98%, TECHNICAL
Barium chloride dihydrate
Formula: BaCl₂·2H₂O
Boiling Pt: 1560 °C (1013 hPa)
Melting Pt: 960 °C
Density: 3,86 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149154
CAS Number: 10326-27-9
EINECS: 233-788-1
UN: 1564
ADR: 6.1,III
Merck Index: 14,00971

Order Now

Specification Test Results

Assay Min. 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR