Barium chloride dihydrate ≥99%, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

VWRC21715.263EA 39.6 SGD
VWRC21715.263 VWRC21715.296
Barium chloride dihydrate ≥99%, GPR RECTAPUR®
Barium chloride dihydrate
Formula: BaCl₂·2H₂O
Boiling Pt: 1560 °C (1013 hPa)
Melting Pt: 960 °C
Density: 3,86 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149154
CAS Number: 10326-27-9
EINECS: 233-788-1
UN: 1564
ADR: 6.1,III
Merck Index: 14,00971

Order Now

Specification Test Results

Assay Min. 99 %
Heavy metals (as Pb) Max. 20 ppm
Insolubility in water Max. 0.05 %
Fe (Iron) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR