Barium chloride dihydrate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent

Supplier: VWR Chemicals
Danger

Synonyms: Barium dichloride

VWRC21716.266EA 111 SGD
VWRC21716.266 VWRC21716.290 VWRC21716.368
Barium chloride dihydrate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Barium chloride dihydrate
Formula: BaCl₂·2H₂O
Boiling Pt: 1560 °C (1013 hPa)
Melting Pt: 960 °C
Density: 3,86 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149154
CAS Number: 10326-27-9
EINECS: 233-788-1
UN: 1564
ADR: 6.1,III
Merck Index: 14,00971

Order Now

Specification Test Results

Assay Min. 99.0 %
pH (25°C; 5 %) 5.2 - 8.2
Heavy metals (as Pb) Max. 5 ppm
Insolubility in water Max. 50 ppm
Loss on drying (150°C) 14.0 - 16.0 %
Total N (Nitrogen) Max. 20 ppm
Ca (Calcium) Max. 50 ppm
Fe (Iron) Max. 1 ppm
K (Potassium) Max. 50 ppm
Na (Sodium) Max. 50 ppm
Pb (Lead) Max. 10 ppm
Sr (Strontium) Max. 0.05 %
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR