Barium sulphate ≥98%, GPR RECTAPUR®

Supplier: VWR Chemicals

VWRC21766.292EA 264 SGD
VWRC21766.292 VWRC21766.463
Barium sulphate ≥98%, GPR RECTAPUR®
Barium sulphate
Formula: BaSO₄
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 1345 °C
Density: 4,5 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003455
CAS Number: 7727-43-7
EINECS: 231-784-4
Merck Index: 13,00997

Order Now

Specification Test Results

Assay Min. 98 %
Acidity Max. 0.002 meq/g
Heavy metals (as Pb) Max. 10 ppm
PO4 (Phosphate) Max. 10 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR