Carboxymethyl cellulose sodium salt, TECHNICAL, medium viscosity

Supplier: VWR Chemicals

Synonyms: Carboxymethyl cellulose, sodium salt, Cellulose gum, Cellulose, carboxymethyl ether, sodium salt, Sodium carboxymethyl cellulose (Viscosity 800mpa.s), Sodium carboxymethyl cellulose, CMC

VWRC22525.296EA 109 SGD
VWRC22525.296
Carboxymethyl cellulose sodium salt, TECHNICAL, medium viscosity
Carboxymethyl cellulose sodium salt
Formula: [C₆H₇O₂(OH)x(OCH₂COONa)y]n
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00081472
CAS Number: 9004-32-4

Order Now

Specification Test Results

Viscosity (25°C; 2 %; water) 300 - 600 mPa.s
Water Max. 8 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR