VWR®, Casein

Supplier: VWR Chemicals

VWRC22544.292EA 314 SGD
VWRC22544.292
VWR®, Casein
Proteins and Peptides

Proteins (on anhydrous product): ≥ 90,0 %
Free acidity: Max. 1,0%
Fats: Max. 2,5%
Ignition residue (SO₄): Max. 2,75%
Loss on drying: Max. 12%

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR