Cobalt(II) chloride hexahydrate 97.0-102.0%, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

VWRC22894.290EA 163 SGD
VWRC22894.290 VWRC22894.184 VWRC22894.261 VWRC22894.465
Cobalt(II) chloride hexahydrate 97.0-102.0%, GPR RECTAPUR®
Cobalt (II) chloride hexahydrate
Formula: CoCl₂·6H₂O
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: 56 °C
Density: 1,92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149652
CAS Number: 7791-13-1
EINECS: 231-589-4
UN: 3077
ADR: 9,III
Merck Index: 13,02462

Order Now

Specification Test Results

Assay (GC) 97.0 - 102.0%
Identification passes test
SO4 (Sulphate) max. 100 ppm
Cu (Copper) max. 50 ppm
Fe (Iron) max. 50 ppm
Ni (Nickel) max. 0.15%
Pb (Lead) max. 30 ppm
Zn (Zinc) max. 0.05%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR