Cobalt(II) chloride hexahydrate 98,0-102,0, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. zA Ni_free

Supplier: VWR Chemicals
Danger

Synonyms: Cobalt Chloride, Hexahydrate, Cobalt dichloride hexahydrate, Cobalt Chloride, 6-Hydrate, Cobaltous chloride hexahydrate

VWRC22896.184EA 40.3 SGD
VWRC22896.184
Cobalt(II) chloride hexahydrate 98,0-102,0, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. zA Ni_free
Cobalt (II) chloride hexahydrate
Formula: CoCl₂·6H₂O
MW: 237.93 g/mol
Boiling Pt: 110
Melting Pt: 56
Density: 1.92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149652
CAS Number: 7791-13-1
EINECS: 231-589-4
UN: 3077
ADR: 9 III
Merck Index: 13,02462

Order Now

Specification Test Results

Assay 98.0 - 102.0 %
Insolubility in water Max. 100 ppm
NO3 (Nitrate) Max. 100 ppm
SO4 (Sulphate) Max. 50 ppm
Ca (Calcium) Max. 50 ppm
Cu (Copper) Max. 5 ppm
Fe (Iron) Max. 10 ppm
K (Potassium) Max. 50 ppm
Mg (Magnesium) Max. 20 ppm
Mn (Manganese) Max. 10 ppm
Na (Sodium) Max. 100 ppm
Ni (Nickel) Max. 50 ppm
Pb (Lead) Max. 5 ppm
Zn (Zinc) Max. 20 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR