Sodium chloride 180 g/l in aqueous solution

Supplier: VWR Chemicals

Synonyms: Saline, NaCl, Sea salt, Halite

VWRC230386MEA 59.2 SGD
VWRC230386M
Sodium chloride 180 g/l in aqueous solution
Sodium chloride

Concentrated saline solution for preparing isotonic (0.9% w/v) solution

Formula: NaCl
Density: 1,2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3

Order Now

Specification Test Results

Assay (W/V) 17.9 - 18.1 %
pH (20°C) 5.5 - 8.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR