Copper(II) sulphate, anhydrous ≥97%, TECHNICAL

Supplier: VWR Chemicals
Warning

Synonyms: Cupric sulphate

VWRC23168.265EA 79.4 SGD
VWRC23168.265
Copper(II) sulphate, anhydrous ≥97%, TECHNICAL
Copper (II) sulphate
Formula: CuSO₄
Melting Pt: 200 °C
Density: 3,603 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010981
CAS Number: 7758-98-7
EINECS: 231-847-6
UN: 3288
ADR: 6.1,III
Merck Index: 12,02722

Order Now

Specification Test Results

Assay Min. 97 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR