Copper(II) sulphate pentahydrate 98,5, purified

Supplier: VWR Chemicals
Warning

Synonyms: Cupric sulphate pentahydrate, Copper sulphate pentahydrate

VWRC23172.293EA 82.6 SGD
VWRC23172.293 VWRC23172.362
Copper(II) sulphate pentahydrate 98,5, purified
Copper (II) sulphate pentahydrate
Formula: CuSO₄·5H₂O
MW: 249.69 g/mol
Density: 2.284 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149681
CAS Number: 7758-99-8
EINECS: 231-847-6
UN: 3077
ADR: 9 III
REACH: 01-2119520566-40
Merck Index: 13,02682

Order Now

Specification Test Results

Assay Min. 98.5 %
Cl (Chloride) Max. 20 ppm
Fe (Iron) Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR