2,6-Dichlorophenolindophenol sodium salt hydrate ACS analytical reagent

Supplier: VWR Chemicals

VWRC23347.105EA 55.7 SGD
VWRC23347.105
2,6-Dichlorophenolindophenol sodium salt hydrate ACS analytical reagent
2,6-Dichlorophenolindophenol sodium salt hydrate
Formula: C₁₂H₆Cl₂NNaO₂·xH₂O
Melting Pt: > 300 °C
MDL Number: MFCD00150014
CAS Number: 1266615-56-8
EINECS: 210-640-4

Order Now

Specification Test Results

IR Spectrum Passes test
Interfering dyestuffs Passes test ACS
Loss on drying (120°C) Max. 12 %
Conforms to ACS Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR