4-Hydroxy-4-methyl-2-pentanone ≥98%, TECHNICAL

Supplier: VWR Chemicals
Warning

Synonyms: Diacetone alcohol

VWRC23424.294EA 186 SGD
VWRC23424.294
4-Hydroxy-4-methyl-2-pentanone ≥98%, TECHNICAL
4-Hydroxy-4-methyl-2-pentanone
Formula: (CH₃)₂C(OH)CH₂COCH₃
Boiling Pt: 166 °C (1013 hPa)
Melting Pt: –44 °C
Density: 0,94 g/cm³ (20 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00004471
CAS Number: 123-42-2
EINECS: 204-626-7
UN: 1148
ADR: 3,III
REACH: 01-2119473975-21
Merck Index: 13,02983
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2da78bff918d252787ee81ab8857bd3b19e51ac&vid=d8a0cccff2cac15b826ba733a013451ac11e3b51&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR