4-Hydroxy-4-methyl-2-pentanone ≥98.0%, GPR RECTAPUR®

Supplier: VWR Chemicals
Warning

Synonyms: Diacetone alcohol

VWRC23427.290EA 251 SGD
VWRC23427.290 VWRC23427.320 VWRC23427.460
4-Hydroxy-4-methyl-2-pentanone ≥98.0%, GPR RECTAPUR®
4-Hydroxy-4-methyl-2-pentanone
Formula: (CH₃)₂C(OH)CH₂COCH₃
Storage Temperature: Ambient
MDL Number: MFCD00004471
CAS Number: 123-42-2
EINECS: 204-626-7
UN: 1148
REACH: 01-2119473975-21

Order Now

Specification Test Results

Assay ≥ 98,0 %
Identification (IR) Passes test Passes test
Colouration ≤ 20 APHA ≤ 20 APHA
Density (20/4) 0,937 - 0,943
Refractive index (20°C) 1,422 - 1,425
Evaporation residue ≤ 0,02 %
Water ≤ 0,1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR