Water, HiPerSolv CHROMANORM® for HPLC

Supplier: VWR Chemicals


Filtered through a 0.2 µm filter.

VWRC23595.294EA 20.9 SGD
VWRC23595.294 VWRC23595.328 VWRC23595.400
Water, HiPerSolv CHROMANORM® for HPLC
Water
Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Filtered at 0,2 µm" Passes test
Colouration Max. 10 APHA
Evaporation residue Max. 1 ppm
Spec. conductivity (25°C)(at filling) Max. 1 µS/cm
Total organic carbon (TOC) Max. 10 ppb
Absorbance (200 nm) Max. 0.01
Absorbance (210 nm) Max. 0.01
Absorbance (220 nm) Max. 0.01
Absorbance (240 nm) Max. 0.01
Absorbance (254 nm) Max. 0.005
Absorbance (280 nm) Max. 0.005
Absorbance (300 nm) Max. 0.005
Fluorescence (as quinine; 254 nm) Max. 1 ppb
Fluorescence (as quinine) (365 nm) Max. 0.5 ppb
Gradient grade (210 nm) Max. 5 mAU
Gradient (254 nm) Max. 1 mAU

SPECIFICATIONS

Environmentally Preferable Safer Human and Environmental Health
Environmental certification EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR