Tin(II) chloride dihydrate ≥95%, TECHNICAL

Supplier: VWR Chemicals
Danger

VWRC23738.232EA 79.8 SGD
VWRC23738.232
Tin(II) chloride dihydrate ≥95%, TECHNICAL
Tin (II) chloride dihydrate
Formula: SnCl₂·2H₂O
Boiling Pt: 623 °C (1013 hPa)
Melting Pt: 246 °C
Density: 2,71 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149863
CAS Number: 10025-69-1
EINECS: 231-868-0
UN: 3260
ADR: 8,III
Merck Index: 13,08861

Order Now

Specification Test Results

Assay Min. 95 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR