Ethyl acetate ≥99%, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: Acetic acid ethyl ester

VWRC23880.290EA 37.7 SGD
VWRC23880.290 VWRC23880.324 VWRC23880.368 VWRC23880.461 VWRC23880.552
Ethyl acetate ≥99%, GPR RECTAPUR®
Ethyl acetate
Formula: CH₃COOC₂H₅
Boiling Pt: 77,1 °C (1013 hPa)
Melting Pt: –83 °C
Density: 0,902 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99 %
IR Spectrum Passes test
Acidity Max. 0.008 meq/g
Boiling point 76.0 - 77.5 °C
Density (20/4) 0.899 - 0.901
Refractive index (20°C) 1.371 - 1.373
Substances coloured by H2SO4 Max. 60 APHA
Ethanol Max. 0.5 %
Evaporation residue Max. 10 ppm
Heavy metals (as Pb) Max. 2 ppm
Water Max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR