Ethyl acetate ≥99.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent

Supplier: VWR Chemicals
Danger

VWRC23882.296EA 36.6 SGD
VWRC23882.296 VWRC23882.321 VWRC23882.467 VWRC23882.550 VWRC23882.330 VWRC23882.310 VWRC23882.360
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Ethyl acetate
Formula: CH₃COOC₂H₅
Boiling Pt: 77,1 °C (1013 hPa)
Melting Pt: –83 °C
Density: 0,902 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.5 %
IR Spectrum Passes test
Organic impurities Passes test
Acidity Max. 0.0006 meq/g
Boiling point 76.0 - 77.5 °C
Colouration Max. 10 APHA
Density (20/4) 0.899 - 0.901
Density (20/20) 0.901 - 0.904
Substances coloured by H2SO4 Max. 60 APHA
Ethanol Max. 0.1 %
Evaporation residue Max. 10 ppm
Methanol Max. 0.1 %
Methyl acetate Max. 0.1 %
iso-Propyl acetate Max. 0.06 %
n-Propyl acetate Max. 0.5 %
Water Max. 0.03 %
Al (Aluminium) Max. 0.1 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.02 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 0.1 ppm
Mg (Magnesium) Max. 0.05 ppm
Mn (Manganese) Max. 0.02 ppm
Na (Sodium) Max. 0.5 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.05 ppm
Sn (Tin) Max. 0.1 ppm
Sr (Strontium) Max. 0.02 ppm
Zn (Zinc) Max. 0.1 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test

SPECIFICATIONS

Environmentally Preferable Safer Human and Environmental Health Safer Human and Environmental Health Safer Human and Environmental Health
Environmental certification Chem21 Solvent Selection Guide recommended by default and after discussion Chem21 Solvent Selection Guide recommended by default and after discussion Chem21 Solvent Selection Guide recommended by default and after discussion

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR