Barium chloride 20% in aqueous solution

Supplier: VWR Chemicals
Warning

VWRC240314.5000EA 437 SGD
VWRC240314.5000
Barium chloride 20% in aqueous solution
Barium chloride
Formula: BaCl₂
Boiling Pt: ∼100 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00003445
CAS Number: 10361-37-2
EINECS: 233-788-1
UN: 3287
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR