Iron(III) chloride 27% in aqueous solution, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: Iron trichloride, Ferric chloride

VWRC24206.297EA 337 SGD
VWRC24206.297
Iron(III) chloride 27% in aqueous solution, GPR RECTAPUR®
Iron (III) chloride
Formula: FeCl₃
MDL Number: MFCD00011005
CAS Number: 7705-08-0
EINECS: 231-729-4
UN: 2582
ADR: 8,III

Order Now

Specification Test Results

Assay 27 - 29 %
Density (20/4) 1.260 - 1.280
Heavy metals (as Pb) Max. 100 ppm
Not precipitated by NH4OH (as SO4) Max. 0.5 %
SO4 (Sulphate) Max. 0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR