Iron(III) chloride hexahydrate 99,0-102,0, AnalaR NORMAPUR® Reag. Ph. Eur., ACS zA

Supplier: VWR Chemicals
Danger

Synonyms: Iron trichloride hexahydrate, Ferric chloride hexahydrate

VWRC24208.237EA 90 SGD
VWRC24208.237 VWRC24208.260 VWRC24208.460
Iron(III) chloride hexahydrate 99,0-102,0, AnalaR NORMAPUR® Reag. Ph. Eur., ACS zA
Iron (III) chloride hexahydrate
Formula: FeCl₃·6H₂O
MW: 270.3 g/mol
Boiling Pt: 280…285 °C (1013 hPa)
Melting Pt: 37
Density: 1.82 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149712
CAS Number: 10025-77-1
EINECS: 231-729-4
UN: 3260
ADR: 8 III
Merck Index: 13,04048
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=808c75376e55f29296d58eb1e5a1e184039ce733&vid=6d820873011bee3ff401441544f7f85599e40626&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0 - 102.0 %
Free chlorine Passes test
Heavy metals (as Pb) Max. 50 ppm
Insoluble substances Max. 100 ppm
Total N (Nitrogen) Max. 10 ppm
Total P (Phosphorus) (as PO4) Max. 100 ppm
NO3 (Nitrate) Max. 100 ppm
SO4 (Sulphate) Max. 100 ppm
Ca (Calcium) Max. 100 ppm
Cu (Copper) Max. 30 ppm
Fe (II) (Iron) Max. 20 ppm
K (Potassium) Max. 50 ppm
Mg (Magnesium) Max. 50 ppm
Na (Sodium) Max. 0.05 %
Zn (Zinc) Max. 30 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR