Iron(III) chloride 41% in aqueous solution, TECHNICAL

Supplier: VWR Chemicals
Danger

Synonyms: Iron trichloride, Ferric chloride

VWRC24212.298EA 72.3 SGD
VWRC24212.298 VWRC24212.367 VWRC24212.447
Iron(III) chloride 41% in aqueous solution, TECHNICAL
Iron (III) chloride
Formula: FeCl₃
MW: 162.21 g/mol
Boiling Pt: 315
Melting Pt: 260
MDL Number: MFCD00011005
CAS Number: 7705-08-0
EINECS: 231-729-4
UN: 2582
ADR: 8 III
Merck Index: 13,04048

Order Now

Specification Test Results

Assay 38 - 42 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR