Iron(II) sulphate hydrate ≥84%, GPR RECTAPUR®

Supplier: VWR Chemicals
Warning

Synonyms: Ferrous sulphate hydrate

VWRC24242.292EA 96.2 SGD
VWRC24242.292
Iron(II) sulphate hydrate ≥84%, GPR RECTAPUR®
Iron (II) sulphate hydrate
Formula: FeSO₄·H₂O
MW: 169.93 g/mol
Melting Pt: ∼300 °C
Density: 2.97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149722
CAS Number: 13463-43-9
EINECS: 231-753-5

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 84 %
Heavy metals (as Pb) Max. 50 ppm
Insolubility in water Max. 0.05 %
As (Arsenic) Max. 2 ppm
Fe (III) (Iron) Max. 0.4 %
Pb (Lead) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR