Iron(III) sulphate hydrate, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: Ferric Persulphate, Diiron trissulphate hydrate, Ferric sulphate hydrate

VWRC24248.292EA 112 SGD
VWRC24248.292
Iron(III) sulphate hydrate, GPR RECTAPUR®
Iron (III) sulphate hydrate
Formula: Fe₂(SO₄)₃·xH₂O
MW: 399.88 g/mol (anhy)
Density: 3.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149714
CAS Number: 15244-10-7
EINECS: 233-072-9
Merck Index: 13,04065

Order Now

Specification Test Results

Assay (calculated as Fe(III)) Min. 21 %
Cl (Chloride) Max. 100 ppm
Fe (II) (Iron) Max. 0.05 %
Zn (Zinc) Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR