D-(-)-Fructose Ph. Eur.

Supplier: VWR Chemicals

VWRC24282.290EA 61.7 SGD
VWRC24282.290 VWRC24282.368 VWRC24282.461
D-(-)-Fructose Ph. Eur.
D(-)-Fructose
Formula: C₆H₁₂O₆
Boiling Pt: 552 °C (1013 hPa)
Melting Pt: 100…110 °C
Density: 1,59 g/cm³ (20 °C. OECD 109)
Storage Temperature: Ambient
MDL Number: MFCD00148910
CAS Number: 57-48-7
EINECS: 200-333-3
Merck Index: 13,04295
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8c96c34b23753d10b06108938876644c747995c&vid=6d952afc3154d3befcf243a015f513a9130c0835&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colourless crystals
Identification A Passes test
Identification B Passes test
Identification C Passes test
Identification D Passes test
Appearance of solution Passes test
Acidity or alkalinity Passes test
Spec. opt. rotation (calc. on anhydrous) -93.5 to -91 deg
Foreign sugars Passes test
5-Hydroxymethylfurfural + related subst. Passes test
Ba (Barium) Passes test
Lead in sugars Max. 0.5 ppm
Water Max. 0.5 %
Sulphated ash Max. 0.1 %
Residual solvents Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR