Hexane (mixture of isomers), TECHNICAL

Supplier: VWR Chemicals
Danger

Synonyms: Hexanes, Isohexane

VWRC24574.323EA 89 SGD
VWRC24574.323 VWRC24574.298 VWRC24574.551 VWRC24574.367 VWRC24574.460
Hexane (mixture of isomers), TECHNICAL
Hexane (mixture of isomers)
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 68…70 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.66 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Boiling point 63 - 69 °C
Evaporation residue Max. 20 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR