n-Hexane 97, HiPerSolv CHROMANORM® HPLC

Supplier: VWR Chemicals
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

Filter 0,2 um_Nitrogen_filled

VWRC24575.320EA 80.2 SGD
VWRC24575.320 VWRC24575.290 VWRC24575.400
n-Hexane 97, HiPerSolv CHROMANORM® HPLC
n-Hexane
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69
Melting Pt: –94,3 °C
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
REACH: 01-2119480412-44
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 97 %
Acidity Max. 0.0003 meq/g
Alkalinity Max. 0.0002 meq/g
Evaporation residue Max. 5 ppm
Water Max. 50 ppm
Absorbance (210 nm) Max. 0.3
Absorbance (220 nm) Max. 0.1
Absorbance (245 nm) Max. 0.01
Transmittance (210 nm) Min. 50 %
Transmittance (220 nm) Min. 80 %
Transmittance (245 nm) Min. 98 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR