n-Hexane 95, AnalaR NORMAPUR® Reag. Ph. Eur., ACS zA

Supplier: VWR Chemicals
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

VWRC24577.298EA 41.1 SGD
VWRC24577.298 VWRC24577.323 VWRC24577.367 VWRC24577.460
n-Hexane 95, AnalaR NORMAPUR® Reag. Ph. Eur., ACS zA
n-Hexane
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69
Melting Pt: –94,3 °C
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
REACH: 01-2119480412-44
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (n-Hexane) Min. 95.0 %
Assay (Isomers) Min. 98.5 %
IR Spectrum Passes test
Thiophene Passes test
Acidity Max. 0.0002 meq/g
Boiling point 68 - 69 °C
Bromine value Max. 0.5
Colouration Max. 10 APHA
Density (20/4) 0.658 - 0.662
Density (20/20) 0.659 - 0.663
Refractive index (20°C) 1.375 - 1.376
Substances coloured by H2SO4 Max. 90 APHA
Benzene Max. 100 ppm
Evaporation residue Max. 10 ppm
Total S (as SO4) Max. 10 ppm
Water Max. 100 ppm
Al (Aluminium) Max. 0.5 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.1 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.01 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.01 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 0.1 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.02 ppm
Na (Sodium) Max. 0.1 ppm
Ni (Nickel) Max. 0.01 ppm
Pb (Lead) Max. 0.01 ppm
Sn (Tin) Max. 0.1 ppm
Sr (Strontium) Max. 0.02 ppm
Zn (Zinc) Max. 0.01 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR