n-Hexane, anhydrous (max. 0.005% H₂O) ≥97%, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

VWRC24603.290EA 136 SGD
VWRC24603.290
n-Hexane, anhydrous (max. 0.005% H₂O) ≥97%, AnalaR NORMAPUR® analytical reagent
n-Hexane
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69
Melting Pt: –94,3 °C
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
REACH: 01-2119480412-44
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colourless liquid
Acidity Max. 0.001 %
Thiophene Passes test
Colour value Max. 10 APHA
Assay Min. 97 %
Residue on evaporation Max. 0.0100 %
Formaldehyde-sulphuric colouration Max. 90 APHA
Benzene Max. 0.01 %
Water Max. 0.005 %
Al (Aluminium) Max. 0.5 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.1 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.01 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.01 ppm
Fe (Iron) Max. 0.1 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.02 ppm
Ni (Nickel) Max. 0.01 ppm
Pb (Lead) Max. 0.01 ppm
Sn (Tin) Max. 0.1 ppm
Zn (Zinc) Max. 0.01 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR