n-Hexane 99, AnalaR NORMAPUR® zA

Supplier: VWR Chemicals
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

VWRC24608.296EA 98.2 SGD
VWRC24608.296 VWRC24608.321 VWRC24608.460
n-Hexane 99, AnalaR NORMAPUR® zA
n-Hexane
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69
Melting Pt: –94,3 °C
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
REACH: 01-2119480412-44
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.0 %
Acidity Max. 0.0004 meq/g
Colouration Max. 10 APHA
Density (20/4) 0.658 - 0.662
Benzene Max. 100 ppm
Evaporation residue Max. 10 ppm
Thiophene Max. 1 ppm
Total S (as SO4) Max. 50 ppm
Water Max. 100 ppm
Al (Aluminium) Max. 0.5 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.1 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.02 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.1 ppm
Sn (Tin) Max. 0.1 ppm
Zn (Zinc) Max. 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR