Hydrazinium sulphate ≥99.0%, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals
Danger

Synonyms: Hydrazine sulphate salt, Hydrazine sulphate

VWRC24696.186EA 76.3 SGD
VWRC24696.186
Hydrazinium sulphate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Hydrazinium sulphate
Formula: NH₂NH₂·H₂SO₄
MW: 130.12 g/mol
Melting Pt: 254
Density: 1.37 g/cm³ (20 °C)
MDL Number: MFCD00044873
CAS Number: 10034-93-2
EINECS: 233-110-4
UN: 3288
ADR: 6.1 III
Merck Index: 14,04772
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e277940036b3cf71839ff7458f0c7d08bff6f6d5&vid=7a677e6f1725cfa1b9bdef05cc04304e6739079e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.0 %
Heavy metals (as Pb) Max. 10 ppm
Ignition residue Max. 0.05 %
Insolubility in water Max. 50 ppm
Cl (Chloride) Max. 50 ppm
Fe (Iron) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR