Lanthanum(III) chloride heptahydrate ≥98%, GPR RECTAPUR®

Supplier: VWR Chemicals

VWRC24957.188EA 62.2 SGD
VWRC24957.188 VWRC24957.268
Lanthanum(III) chloride heptahydrate ≥98%, GPR RECTAPUR®
Lanthanum (III) chloride heptahydrate
Formula: LaCl₃·7H₂O
Boiling Pt: < 860 °C (1013 hPa)
Melting Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00149756
CAS Number: 10025-84-0
EINECS: 233-237-5

Order Now

Specification Test Results

Assay Min. 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR