D-(-)-Fructose, GPR RECTAPUR®

Supplier: VWR Chemicals

Synonyms: D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose

VWRC24975.294EA 54.4 SGD
VWRC24975.294
D-(-)-Fructose, GPR RECTAPUR®
D(-)-Fructose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 552
Melting Pt: 100…110 °C
Density: 1.59 g/cm³ (20 °C. OECD 109)
Storage Temperature: Ambient
MDL Number: MFCD00148910
CAS Number: 57-48-7
EINECS: 200-333-3
Merck Index: 13,04295
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8c96c34b23753d10b06108938876644c747995c&vid=6d952afc3154d3befcf243a015f513a9130c0835&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Specific optical rotation (10 %; water) -94 to -90 °
Ignition residue (SO4) Max. 0.1 %
Water Max. 0.5 %
Cl (Chloride) Max. 100 ppm
SO4 (Sulphate) Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR