Peqlab peqGOLD, DNA Ladder, 100 bp Plus
Supplier: VWR Chemicals
VWRC25-2020EA 266 SGD
VWRC25-2020
Peqlab peqGOLD, DNA Ladder, 100 bp Plus
Electrophoresis Reagents Molecular Weight Markers
peqGOLD DNA ladder 100 bp plus is ideal for size determination in the range of 100 to 3000 base pairs. There are no unspecific bands besides the fragments described. 1000 and 500 bp bands have a higher DNA content and serve as a size reference.
  • Composed of 14 chromatography-purified individual DNA fragments: 3000, 2000, 1500, 1200, 1000, 900, 800, 700, 600, 500, 400, 300, 200 and 100 base pairs
Delivery information: Product comprises of 100 µl (50 µg) DNA ladder and 1 ml 6X loading buffer (10 mM Tris-HCl, pH 7,6; 0,03% bromophenol blue; 0,03% xylene cyanol; 60% glycerol and 60 mM EDTA)
Caution: The marker has to be dephosphorylated before being used in radioactive assays.
Order Now
SPECIFICATIONS
Environmentally Preferable