Lithium chloride 99,0, AnalaR NORMAPUR® Reag. Ph. Eur. zA

Supplier: VWR Chemicals
Warning

VWRC25012.237EA 147 SGD
VWRC25012.237 VWRC25012.363 VWRC25012.260 VWRC25012.410
Lithium chloride 99,0, AnalaR NORMAPUR® Reag. Ph. Eur. zA
Lithium chloride
Formula: LiCl
MW: 42.39 g/mol
Boiling Pt: 1382
Melting Pt: 614
Density: 2.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3
Merck Index: 13,05550

Order Now

Specification Test Results

Assay Min. 99.0 %
pH (20°C; 5 %) 5 - 9
Heavy metals (as Pb) Max. 5 ppm
Total N (Nitrogen) Max. 10 ppm
SO4 (Sulphate) Max. 50 ppm
Ba (Barium) Max. 20 ppm
Ca (Calcium) Max. 50 ppm
Fe (Iron) Max. 5 ppm
K (Potassium) Max. 0.05 %
Mg (Magnesium) Max. 50 ppm
Na (Sodium) Max. 0.05 %
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR