VWR® Contact Plates, Blister Packed, Irradiated
Supplier: VWR Chemicals
VWRC251183ZAEA 98.8 SGD
VWRC251183ZA VWRC250641ZA VWRC250063UA VWRC250544ZI VWRC251114ZI VWRC271114ZI VWRC251114PY VWRC251114RY VWRC271114RY VWRC251114TI VWRC271114TI VWRC250884ZI VWRC270884ZI VWRC250884TI VWRC270884TI VWRC250774ZI VWRC270774ZI VWRC250774TI VWRC270774TI VWRC250070ZI VWRC251203ZA VWRC250793ZI VWRC250884CY VWRC250560ZA VWRC251114SI VWRC251114AF VWRC251203TA VWRC250542CA VWRC251114SIMP VWRC251114RYMP VWRC251114PYMP VWRC251114AFMP VWRC250884CYMP VWRC250063UAMP VWRC250070ZIMP VWRC251114ZIMP VWRC251114TIMP VWRC251203TAMP VWRC251183ZAMP VWRC270884CY
VWR® Contact Plates, Blister Packed, Irradiated
Media for Microbiology Contact Plates for Microbiology
Available in two pack sizes - triple-wrapped (24 plate pack) and double-wrapped (30 plate pack). Plates are packed in a pack with six blisters. Each blister contains one plate, so every time you open a compartment you use only one plate. Available with TLHTh disinfectant neutraliser (Tween®, lecithin, histidine, thiosulphate).
  • Triple-wrapping ensures that the package itself doesn't contaminate the environment, as the first wrapper is removed just before entering a clean area
  • Available with certificate of irradiation/dosimetry
  • VHP resistance studies available for all triple-wrapped plates
  • Storage at room temperature
Packaging: 5×6 or 4×5×6 plate packs consist of 5 blister packs with an additional cellophane bag. 4×6 plate packs comprise of 4 blister packs with 2 additional cellophane bags. An irradiation indicator is featured on each blister.