Tin(II) chloride dihydrate ≥98%, AnalaR NORMAPUR® analytical reagent (max. 0.05 ppm Hg)

Supplier: VWR Chemicals
Danger

Synonyms: Stannous chloride dihydrate

VWRC2512.0250EA 79.4 SGD
VWRC2512.0250
Tin(II) chloride dihydrate ≥98%, AnalaR NORMAPUR® analytical reagent (max. 0.05 ppm Hg)
Tin (II) chloride dihydrate
Formula: SnCl₂·2H₂O
Boiling Pt: 623 °C (1013 hPa)
Melting Pt: 246 °C
Density: 2,71 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149863
CAS Number: 10025-69-1
EINECS: 231-868-0
UN: 3260
ADR: 8,III
Merck Index: 13,08861

Order Now

Specification Test Results

Assay Min. 98 %
Insolubility in diluted HCl Passes test
NH4 (Ammonium) Max. 20 ppm
SO4 (Sulphate) Max. 20 ppm
As (Arsenic) Max. 1 ppm
Ca (Calcium) Max. 0,005%
Cu (Copper) Max. 10 ppm
Fe (Iron) Max. 20 ppm
Hg (Mercury) Max. 0.05 ppm
K (Potassium) Max. 50 ppm
Na (Sodium) Max. 100 ppm
Pb (Lead) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR