Magnesium sulphate heptahydrate ≥98%, purified

Supplier: VWR Chemicals

VWRC25163.364EA 76.2 SGD
VWRC25163.364 VWRC25163.290 VWRC25163.460
Magnesium sulphate heptahydrate ≥98%, purified
Magnesium sulphate heptahydrate
Formula: MgSO₄·7H₂O
Melting Pt: 1124 °C
Density: 2,66 g/cm³ (20 °C)
MDL Number: MFCD00149785
CAS Number: 10034-99-8
Merck Index: 13,05715

Order Now

Specification Test Results

Assay Min. 98 %
Insoluble matter Max. 0.05 %
Fe (Iron) Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR