Methyl-4-hydroxybenzoate ≥99%, TECHNICAL

Supplier: VWR Chemicals
Warning

VWRC25605.293EA 203 SGD
VWRC25605.293
Methyl-4-hydroxybenzoate ≥99%, TECHNICAL
Methyl 4-hydroxybenzoate
Formula: HOC₆H₄CO₂CH₃
Boiling Pt: 270…280 °C (1013 hPa)
Melting Pt: 125…128 °C
Density: 1,1208 g/cm³ (137 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002352
CAS Number: 99-76-3
EINECS: 202-785-7
Merck Index: 13,06129
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84321052052f4be2a53d8d7ebac7fabe6fc5a435&vid=ede765fad275b2515a8c57ba4499bff313a18983&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR