Methyl ethyl ketone ≥99%, TECHNICAL

Supplier: VWR Chemicals
Danger

VWRC25654.292EA 62.4 SGD
VWRC25654.292 VWRC25654.361 VWRC25654.320
Methyl ethyl ketone ≥99%, TECHNICAL
Methyl ethyl ketone
Formula: C₂H₅COCH₃
Boiling Pt: 79,6 °C (1013 hPa)
Melting Pt: –86 °C
Density: 0,8054 g/cm³ (20 °C)
Flash Pt: –4 °C (DIN 51755 Part 1)
Storage Temperature: Ambient
MDL Number: MFCD00011648
CAS Number: 78-93-3
EINECS: 201-159-0
UN: 1193
ADR: 3,II
REACH: 01-2119457290-43
Merck Index: 13,06097
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=17cb98abf677b90f8bac752664d5c0cfd6765591&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99 %
Boiling point 79 - 80 °C
Density (20/4) 0.804 - 0.807


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR